Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glyoxime ≥98%

Supplier: Thermo Scientific Chemicals
Danger

L05130-09 L05130-18 L05130-18 L05130-09
AAAL05130-09EA 71.25 USD InStock
AAAL05130-09 CAAAAL05130-18 CAAAAL05130-09 AAAL05130-18
Glyoxime ≥98%
Glyoxime


Formula: C₂H₄N₂O₂
Melting Pt: ca 169 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013942
CAS Number: 557-30-2
EINECS: 209-168-1
UN: 2926
ADR: "4.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty