Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-α,α-dimethylphenylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(4-Chlorophenyl)-2-methylpropanoic acid

L05277-03 L05277-06 L05277-03 L05277-06
AAAL05277-03EA 41.09 USD InStock
CAAAAL05277-03 AAAL05277-03 CAAAAL05277-06 AAAL05277-06
4-Chloro-α,α-dimethylphenylacetic acid 98%
4-Chloro-alpha,alpha-dimethylphenylacetic acid


Formula: C₁₀H₁₁ClO₂
MW: 198.65 g/mol
Melting Pt: 124…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00044814
CAS Number: 6258-30-6
EINECS: 228-392-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty