Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-[4-(Acetylamino)phenyl]acrylic acid predominantly trans 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: trans-4-(Acetylamino)cinnamic acid

L05292-09 L05292-09 L05292-04 L05292-04
AAAL05292-04EA 105.99 USD InStock
AAAL05292-04 CAAAAL05292-04 AAAL05292-09 CAAAAL05292-09
3-[4-(Acetylamino)phenyl]acrylic acid predominantly trans 98%
3-[4-(Acetylamino)phenyl]acrylic acid


Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
Melting Pt: ca 259 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016846
CAS Number: 7152-04-7
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty