Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Phenyl-1-(trimethylsilyloxy)ethylene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Trimethyl[(1-phenylvinyl)oxy]silane1-Phenyl-1-trimethylsiloxyethylene

L05308-03 L05308-06 L05308-03 L05308-06
AAAL05308-06EA 100.99 USD InStock
AAAL05308-06 CAAAAL05308-03 CAAAAL05308-06 AAAL05308-03
1-Phenyl-1-(trimethylsilyloxy)ethylene 97%
1-Phenyl-1-(trimethylsilyloxy)ethylene


Formula: C₁₁H₁₆OSi
MW: 192.34 g/mol
Boiling Pt: 53…54 °C/1 mm
Density: 0,938
Flash Pt: 78 °C(172 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008582
CAS Number: 13735-81-4
EINECS: 237-308-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty