Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorobenzylideneacetone 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(4-Chlorophenyl)but-3-en-2-one

L05321-06 L05321-14 L05321-14 L05321-06
AAAL05321-06EA 51.69 USD InStock
AAAL05321-06 CAAAAL05321-14 AAAL05321-14 CAAAAL05321-06
4-Chlorobenzylideneacetone 98%
4-Chlorobenzylideneacetone


Formula: C₁₀H₉ClO
MW: 180.64 g/mol
Boiling Pt: 145…147 °C/5 mm
Melting Pt: 55…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00018790
CAS Number: 3160-40-5
EINECS: 221-610-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty