Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Triacontane 99%

Supplier: Thermo Scientific Chemicals

L05386-03 L05386-06 L05386-03 L05386-MD L05386-06 L05386-MD
AAAL05386-03EA 62.19 USD InStock
AAAL05386-03 CAAAAL05386-03 AAAL05386-MD AAAL05386-06 CAAAAL05386-MD CAAAAL05386-06
n-Triacontane 99%
n-Triacontane


Formula: H₃C(CH₂)₂₈CH₃
MW: 422.82 g/mol
Boiling Pt: 455 °C
Melting Pt: 65…68 °C
Density: 0.7797 g/cm³ (20 °C)
Flash Pt: 237 °C(458 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009410
CAS Number: 638-68-6
EINECS: 211-349-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty