Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitro-m-xylene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,6-Dimethylnitrobenzene2-Nitro-m-xylene1,3-Dimethyl-2-nitrobenzene

L05546-36 L05546-36 L05546-22 L05546-22
AAAL05546-36EA 136.99 USD InStock
AAAL05546-36 CAAAAL05546-22 CAAAAL05546-36 AAAL05546-22
2-Nitro-m-xylene 99%
2-Nitro-m-xylene


Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 225…227 °C
Melting Pt: 14…16 °C
Density: 1.110
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007181
CAS Number: 81-20-9
EINECS: 201-333-6
UN: 1665
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty