Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentyl-4-hydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Pentylparaben4-Hydroxybenzoic acid pentyl esterPentyl parahydroxybenzoate

L05563-14 L05563-14 L05563-06 L05563-06
AAAL05563-06EA 47.19 USD InStock
AAAL05563-06 CAAAAL05563-06 AAAL05563-14 CAAAAL05563-14
Pentyl-4-hydroxybenzoate 98%
Pentyl 4-hydroxybenzoate


Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
Boiling Pt: 197…200 °C/10 mm
Melting Pt: 36…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00016479
CAS Number: 6521-29-5
EINECS: 229-408-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty