Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Octyn-1-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

L05596-18 L05596-09 L05596-09 L05596-18
AAAL05596-18EA 323.99 USD InStock
CAAAAL05596-09 AAAL05596-18 CAAAAL05596-18 AAAL05596-09
2-Octyn-1-ol 98%
2-Octyn-1-ol


Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 76…78 °C/2 mm
Density: 0.880
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039542
CAS Number: 20739-58-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty