Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(2-Aminoethyl)pyridine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(2-Pyridyl)ethylamine2-Pyridylethylamine2-Pyridin-2-ylethanamine

L05627-06 L05627-03 L05627-14 L05627-14 L05627-06 L05627-03
AAAL05627-14EA 327.99 USD InStock
CAAAAL05627-06 AAAL05627-14 AAAL05627-03 CAAAAL05627-03 CAAAAL05627-14 AAAL05627-06
2-(2-Aminoethyl)pyridine 98%
2-(2-Aminoethyl)pyridine


Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 208 °C
Density: 1.020
Flash Pt: 100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006367
CAS Number: 2706-56-1
EINECS: 220-295-1
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty