Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Morpholinoaniline 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(4-Morpholinyl)aniline

L05660-06 L05660-14 L05660-06 L05660-14
AAAL05660-06EA 85.59 USD InStock
CAAAAL05660-14 AAAL05660-06 CAAAAL05660-06 AAAL05660-14
2-Morpholinoaniline 98%
2-Morpholinoaniline


Formula: C₁₀H₁₄N₂O
MW: 178.23 g/mol
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00047408
CAS Number: 5585-33-1
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty