Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-1-Phenyl-1-propanol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-(+)-α-Ethylbenzyl alcohol

L05681-MC L05681-ME L05681-ME L05681-MC
AAAL05681-MEEA 150.99 USD InStock
AAAL05681-ME CAAAAL05681-ME AAAL05681-MC CAAAAL05681-MC
(R)-(+)-1-Phenyl-1-propanol 99%
(R)-(+)-1-Phenyl-1-propanol
For synthesis of optically active products


Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 217…219 °C
Density: 0.993
Flash Pt: 95 °C(203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064279
CAS Number: 1565-74-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty