Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chlorothiazol-2-amine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

L05707-03 L05707-06 L05707-14 L05707-03 L05707-14 L05707-06
AAAL05707-06EA 100.99 USD InStock
AAAL05707-06 AAAL05707-03 AAAL05707-14 CAAAAL05707-06 CAAAAL05707-03 CAAAAL05707-14
5-Chlorothiazol-2-amine hydrochloride 97%
5-Chlorothiazol-2-amine hydrochloride


Formula: C₃H₄Cl₂N₂S
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00039006
CAS Number: 55506-37-1
EINECS: 259-680-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty