Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Ethoxybenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

L05721-09 L05721-09
AAAL05721-09EA 105.99 USD InStock
CAAAAL05721-09 AAAL05721-09
3-Ethoxybenzoic acid 98+%
3-Ethoxybenzoic acid


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00016517
CAS Number: 621-51-2
EINECS: 210-690-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty