Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethyl)benzoic acid hydrazide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Trifluoromethyl)benzohydrazide4-(Trifluoromethyl)benzoic Hydrazide4-(Trifluoromethyl)benzhydrazide4-(Trifluoromethyl)benzoylhydrazine

L05724-03 L05724-06 L05724-06 L05724-03
AAAL05724-06EA 151.99 USD InStock
CAAAAL05724-06 AAAL05724-06 CAAAAL05724-03 AAAL05724-03
4-(Trifluoromethyl)benzoic acid hydrazide 98%
4-(Trifluoromethyl)benzoic acid hydrazide


Formula: C₈H₇F₃N₂O
MW: 204.15 g/mol
Melting Pt: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00051703
CAS Number: 339-59-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty