Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Chloroethyl)morpholine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-(2-Chloroethyl)-Morpholinium ChlorideN-(2-Chloroethyl)morpholine hydrochloride

L05771-36 L05771-22 L05771-22 L05771-36
AAAL05771-36EA 121.99 USD InStock
AAAL05771-36 AAAL05771-22 CAAAAL05771-36 CAAAAL05771-22
4-(2-Chloroethyl)morpholine hydrochloride 99%
4-(2-Chloroethyl)morpholine hydrochloride


Formula: C₆H₁₃Cl₂NO
MW: 186.08 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00012797
CAS Number: 3647-69-6
EINECS: 222-881-2
UN: 2923
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty