Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Benzylcyclohexanone 97%

Supplier: Thermo Scientific Chemicals
Warning

L05776-03 L05776-03 L05776-06 L05776-06
AAAL05776-03EA 42.39 USD InStock
CAAAAL05776-03 AAAL05776-03 CAAAAL05776-06 AAAL05776-06
2-Benzylcyclohexanone 97%
2-Benzylcyclohexanone


Formula: C₁₃H₁₆O
MW: 188.27
Boiling Pt: 103…105 °C/0.2 mm
Melting Pt: 28…30 °C
Density: 1.030
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051455
EINECS: 213-420-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty