Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Mercapto-4-phenylthiazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Phenylthiazole-2-thiol

L05829-06 L05829-14 L05829-14 L05829-06
AAAL05829-14EA 300.99 USD InStock
AAAL05829-14 CAAAAL05829-06 AAAL05829-06 CAAAAL05829-14
2-Mercapto-4-phenylthiazole 98%
2-Mercapto-4-phenylthiazole


Formula: C₉H₇NS₂
MW: 193.30 g/mol
Melting Pt: 172…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00014508
CAS Number: 2103-88-0
EINECS: 218-274-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty