Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-2-Decenal 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (E)-2-Decenal(E)-Dec-2-enal

L05901-14 L05901-06 L05901-06 L05901-14
AAAL05901-14EA 218.99 USD InStock
CAAAAL05901-14 CAAAAL05901-06 AAAL05901-14 AAAL05901-06
trans-2-Decenal 95%
trans-2-Decenal


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 117 °C/17 mm
Density: 0.840
Flash Pt: 93 °C(199 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00014679
CAS Number: 3913-81-3
EINECS: 223-474-2
UN: 3082
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty