Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Margaric acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Heptadecanoic acidHeptadecylic acid

L05928-06 L05928-14 L05928-14 L05928-06
AAAL05928-06EA 48.69 USD InStock
AAAL05928-06 AAAL05928-14 CAAAAL05928-06 CAAAAL05928-14
Margaric acid 98%
Margaric acid


Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 227 °C/100 mm
Melting Pt: 59…63 °C
Density: 0,853
Storage Temperature: Ambient
MDL Number: MFCD00002751
CAS Number: 506-12-7
EINECS: 208-027-1
Merck Index: 13,05775
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty