Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Aminobutyric acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-DL-Abu-OHDL-2-Aminobutyric acidDL-α-Amino-N-Butyric Acid

L06035-14 L06035-14 L06035-22 L06035-22
AAAL06035-22EA 115.99 USD InStock
AAAL06035-22 CAAAAL06035-14 AAAL06035-14 CAAAAL06035-22
(±)-2-Aminobutyric acid 99%
(±)-2-Aminobutyric acid


Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: ca 291 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008093
CAS Number: 2835-81-6
EINECS: 220-616-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty