Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Tritylphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

L06073-06 L06073-14 L06073-06 L06073-14
AAAL06073-14EA 147.99 USD InStock
CAAAAL06073-14 CAAAAL06073-06 AAAL06073-14 AAAL06073-06
4-Tritylphenol 98%
4-Tritylphenol


Formula: C₂₅H₂₀O
Melting Pt: 276…286 °C
Storage Temperature: Ambient
MDL Number: MFCD00002364
CAS Number: 978-86-9
EINECS: 213-557-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty