Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxybenzoic anhydride 98%

Supplier: Thermo Scientific Chemicals
Warning

L06180-14 L06180-14 L06180-06 L06180-06
AAAL06180-14EA 262.99 USD InStock
CAAAAL06180-14 CAAAAL06180-06 AAAL06180-14 AAAL06180-06
4-Methoxybenzoic anhydride 98%
4-Methoxybenzoic anhydride


Formula: C₁₆H₁₄O₅
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00017175
CAS Number: 794-94-5
EINECS: 212-345-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty