Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L06240-18 L06240-30 L06240-09 L06240-09 L06240-30 L06240-18
AAAL06240-18EA 90.09 USD InStock
CAAAAL06240-18 AAAL06240-18 CAAAAL06240-09 AAAL06240-09 CAAAAL06240-30 AAAL06240-30
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid 98%
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid


Formula: C₁₁H₈ClNO₃
Melting Pt: 187…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00020813
CAS Number: 23598-72-3
EINECS: 245-770-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty