Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-1-methylpiperidine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Methylpiperidin-4-ol1-Methyl-4-piperidinolN-Methyl-4-piperidinol

L06415-36 L06415-22 L06415-36 L06415-22
AAAL06415-22EA 52.99 USD InStock
CAAAAL06415-36 AAAL06415-22 CAAAAL06415-22 AAAL06415-36
4-Hydroxy-1-methylpiperidine 98%
4-Hydroxy-1-methylpiperidine


Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 198…200 °C
Melting Pt: 26…30 °C
Density: 0,98
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006500
CAS Number: 106-52-5
EINECS: 203-406-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty