Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Nitro-o-toluic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Methyl-6-nitrobenzoic acid

L06563-03 L06563-06 L06563-03 L06563-14 L06563-14 L06563-06
AAAL06563-03EA 27.36 USD InStock
AAAL06563-03 AAAL06563-14 CAAAAL06563-03 CAAAAL06563-14 CAAAAL06563-06 AAAL06563-06
6-Nitro-o-toluic acid 98%
6-Nitro-o-toluic acid


Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 353 °C (1013 hPa)
Melting Pt: 153…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007267
CAS Number: 13506-76-8
EINECS: 236-833-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty