Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

L06831-14 L06831-14 L06831-06 L06831-22 L06831-22 L06831-06
AAAL06831-22EA 201.99 USD InStock
CAAAAL06831-22 CAAAAL06831-14 AAAL06831-22 AAAL06831-14 CAAAAL06831-06 AAAL06831-06
4-Fluorobenzamide 98%
4-Fluorobenzamide


Formula: C₇H₆FNO
MW: 139.13 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 154…157 °C
Density: 1,43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007992
CAS Number: 824-75-9
EINECS: 212-535-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty