Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disoprofol 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,6-BisisopropylphenolPropofol2,6-Diisopropylphenol1,3-Di(methylethyl)-2-hydroxybenzene

L06841-22 L06841-14 L06841-36 L06841-22 L06841-36 L06841-14
AAAL06841-14EA 47 USD InStock
AAAL06841-14 AAAL06841-36 CAAAAL06841-36 CAAAAL06841-14 AAAL06841-22 CAAAAL06841-22
Disoprofol 97%
Disoprofol
An anesthetic that exhibits direct activation of the GABA-A receptor


Formula: C₁₂H₁₈O
Boiling Pt: 254…256 °C
Melting Pt: 17…18 °C
Density: 0.962
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008885
CAS Number: 2078-54-8
EINECS: 218-206-6
UN: 3145
ADR: "8",III
Merck Index: 13,07927
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty