Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,2-Trichloroethyl chloroformate 97%

Supplier: Thermo Scientific Chemicals
Danger

L06875-14 L06875-22 L06875-22 L06875-14
AAAL06875-14EA 51.19 USD InStock
AAAL06875-14 CAAAAL06875-14 AAAL06875-22 CAAAAL06875-22
2,2,2-Trichloroethyl chloroformate 97%
2,2,2-Trichloroethyl chloroformate


Formula: C₃H₂Cl₄O₂
MW: 211.86 g/mol
Boiling Pt: 171…172 °C
Melting Pt: 0…2 °C
Density: 1.539
Flash Pt: >100 °C(212 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000810
CAS Number: 17341-93-4
EINECS: 241-363-7
UN: 3277
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty