Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-6-chlorobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

L06883-03 L06883-03 L06883-06 L06883-06
AAAL06883-03EA 28.19 USD InStock
CAAAAL06883-03 AAAL06883-03 CAAAAL06883-06 AAAL06883-06
2-Amino-6-chlorobenzonitrile 98%
2-Amino-6-chlorobenzonitrile


Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00015546
CAS Number: 6575-11-7
EINECS: 229-501-4
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty