Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-3-nitroanisole 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Bromo-4-methoxy-2-nitrobenzene

L06944-06 L06944-14 L06944-06 L06944-14
AAAL06944-14EA 94.09 USD InStock
CAAAAL06944-14 CAAAAL06944-06 AAAL06944-14 AAAL06944-06
4-Bromo-3-nitroanisole 96%
4-Bromo-3-nitroanisole


Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Boiling Pt: 153…154 °C/13 mm
Melting Pt: 29…32 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051511
CAS Number: 5344-78-5
EINECS: 226-290-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty