Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloroanisole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Chloro-2-methoxybenzene

L06952-30 L06952-18 L06952-30 L06952-18
AAAL06952-30EA 176.99 USD InStock
CAAAAL06952-30 AAAL06952-30 AAAL06952-18 CAAAAL06952-18
2-Chloroanisole 98%
2-Chloroanisole


Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 194…196 °C
Melting Pt: –27 °C
Density: 1,123
Flash Pt: 76 °C(168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000557
CAS Number: 766-51-8
EINECS: 212-167-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty