Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Chloromethyl)cyclopropane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cyclopropyl methyl chloride

L06986-14 L06986-14 L06986-06
AAAL06986-14EA 315.99 USD InStock
AAAL06986-14 CAAAAL06986-06 CAAAAL06986-14
(Chloromethyl)cyclopropane 97%
(Chloromethyl)cyclopropane


Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 87…89 °C
Melting Pt: –91 °C
Density: 0.980
Flash Pt: -1 °C(30 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001307
CAS Number: 5911-08-0
EINECS: 227-632-1
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty