Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Dodecylmethylamine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-methyldodecylamine

L07025-03 L07025-06 L07025-06
AAAL07025-06EA 163.99 USD InStock
CAAAAL07025-06 AAAL07025-06 AAL07025-03
N-Dodecylmethylamine 98%
N-Dodecylmethylamine


Formula: CH₃(CH₂)₁₁NHCH₃
MW: 199.38 g/mol
Boiling Pt: 90…92 °C/0.9 mm
Density: 0.795
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026574
CAS Number: 7311-30-0
EINECS: 230-771-0
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty