Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromopyridinium chloride 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Bromopyridine hydrochloride4-Bromopyridine HCl

L07052-03 L07052-06 L07052-14 L07052-14 L07052-03 L07052-06
AAAL07052-06EA 103.99 USD InStock
AAAL07052-06 AAAL07052-14 AAAL07052-03 CAAAAL07052-06 CAAAAL07052-03 CAAAAL07052-14
4-Bromopyridinium chloride 99%
4-Bromopyridinium chloride


Formula: C₅H₅BrClN
MW: 194.46 g/mol
Melting Pt: ca 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty