Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-α-(4-Hydroxyphenyl)glycine 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-D-Phg(4-OH)-OH(-)-4-Hydroxy-D-phenylglycine(R)-(-)-2-(4-Hydroxyphenyl)glycineD-(-)-p-HydroxyphenylglycineD(-)-4-Hydroxyphenylglycine

L07190-14 L07190-14 L07190-06 L07190-06
AAAL07190-14EA 43.99 USD InStock
CAAAAL07190-14 CAAAAL07190-06 AAAL07190-14 AAAL07190-06
D-(-)-α-(4-Hydroxyphenyl)glycine 98+%
D(-)-α-(4-Hydroxyphenyl)glycine


Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Ambient
EINECS: 245-247-7
Order Now
Specification Test Results
Optical rotation -156° (c=1 in 1N HCl)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty