Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethyl-9,10-anthraquinone 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Ethylanthracene-9,10-dione2-Ethylanthraquinone

L07206-06 L07206-36 L07206-22 L07206-36 L07206-06 L07206-22
AAAL07206-22EA 43.09 USD InStock
AAAL07206-22 CAAAAL07206-22 AAAL07206-36 CAAAAL07206-06 CAAAAL07206-36 AAAL07206-06
2-Ethyl-9,10-anthraquinone 98%
2-Ethyl-9,10-anthraquinone


Formula: C₁₆H₁₂O₂
MW: 236.27 g/mol
Boiling Pt: 180…190 °C
Melting Pt: 107…111 °C
Flash Pt: >210 °C(410 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001237
CAS Number: 84-51-5
EINECS: 201-535-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty