Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-tert-Butylphenol 99%

Supplier: Thermo Scientific Chemicals
Danger

L07294-22 L07294-0P L07294-0P L07294-22
AAAL07294-0PEA 107.99 USD InStock
CAAAAL07294-22 CAAAAL07294-0P AAAL07294-0P AAAL07294-22
2-tert-Butylphenol 99%
2-tert-Butylphenol


Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 223…224 °C
Melting Pt: –7 °C
Density: 0.980
Flash Pt: 102 °C(215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002223
CAS Number: 88-18-6
EINECS: 201-807-2
UN: 2922
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty