Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Deoxy-D-glucose 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal

L07338-14 L07338-03 L07338-06 L07338-06 L07338-14 L07338-03
AAAL07338-14EA 848.99 USD InStock
AAAL07338-14 AAAL07338-03 CAAAAL07338-06 CAAAAL07338-14 CAAAAL07338-03 AAAL07338-06
2-Deoxy-D-glucose 98%
2-Deoxy-D-glucose
A useful culture media component for molecular genetics


Formula: C₆H₁₂O₅
MW: 164.16 g/mol
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00151328
CAS Number: 154-17-6
EINECS: 205-823-0
Order Now
Specification Test Results
Optical rotation +47° (c=0.5 in water, 45h)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty