Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Methylthio)-m-cresol 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Methyl-4-(methylthio)phenol

L07353-06 L07353-14 L07353-06 L07353-14
AAAL07353-06EA 38.49 USD InStock
CAAAAL07353-14 AAAL07353-06 AAAL07353-14 CAAAAL07353-06
4-(Methylthio)-m-cresol 97%
4-(Methylthio)-m-cresol


Formula: C₈H₁₀OS
MW: 154.23 g/mol
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00045773
CAS Number: 3120-74-9
EINECS: 221-496-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty