Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Methyltetrahydrofuran 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) ∼ 150-400 ppm

L07356-0F L07356-AE L07356-AP L07356-AP L07356-0F L07356-AE
AAL07356-0FEA 542.43 USD InStock
CAAAAL07356-AE AAL07356-0F AAAL07356-AE CAAAAL07356-AP AAAL07356-AP CAAAL07356-0F
(±)-2-Methyltetrahydrofuran 99% stabilized
(±)-2-Methyltetrahydrofuran


Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 80…82 °C
Melting Pt: –136 °C
Density: 0.856
Flash Pt: -11 °C(12 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: "3",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty