Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzanthrone ≥85%, tech.

Supplier: Thermo Scientific Chemicals
Danger

L07422-22 L07422-14 L07422-14 L07422-22
AAAL07422-14EA 62.59 USD InStock
AAAL07422-14 CAAAAL07422-14 AAAL07422-22 CAAAAL07422-22
3-Bromobenzanthrone ≥85%, tech.
3-Bromobenzanthrone


Formula: C₁₇H₉BrO
MW: 309.17 g/mol
Melting Pt: 164…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00021094
CAS Number: 81-96-9
EINECS: 201-390-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty