Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-6-hydroxy-2-methylpyrimidine dihydrate ≥98%

Supplier: Thermo Scientific Chemicals

L07496-06 L07496-06 L07496-14 L07496-14
AAAL07496-14EA 127.99 USD InStock
CAAAAL07496-14 CAAAAL07496-06 AAAL07496-14 AAAL07496-06
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate ≥98%
4-Amino-6-hydroxy-2-methylpyrimidine dihydrate


Formula: C₅H₇N₃O·2H₂O
MW: 161.16 g/mol (125.13 anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150311
CAS Number: 388582-41-0
EINECS: 212-182-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty