Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N(α)-Cbz-L-tryptophan 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: Z-Trp-OHN-Cbz-L-TryptophanN(α)-Benzyloxycarbonyl-L-tryptophan(S)-2-(((Benzyloxy)carbonyl)amino)-3-(1H-indol-3-yl)propanoic acid

L07535-14 L07535-06 L07535-06 L07535-14
AAAL07535-14EA 78.09 USD InStock
AAAL07535-14 CAAAAL07535-14 AAAL07535-06 CAAAAL07535-06
N(α)-Cbz-L-tryptophan 98+%
N(α)-Cbz-L-tryptophan


Formula: C₁₉H₁₈N₂O₄
MW: 338.4 g/mol
Melting Pt: 121…126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065700
CAS Number: 7432-21-5
EINECS: 231-074-4
Order Now
Specification Test Results
Optical rotation +3.1° (c=3 in acetic acid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty