Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-chloro-4-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

L07573-18 L07573-09 L07573-09 L07573-18
AAAL07573-18EA 241.99 USD InStock
CAAAAL07573-18 AAAL07573-18 AAAL07573-09 CAAAAL07573-09
1-Bromo-2-chloro-4-nitrobenzene 98%
1-Bromo-2-chloro-4-nitrobenzene


Formula: C₆H₃BrClNO₂
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00051514
CAS Number: 29682-39-1
EINECS: 249-773-8
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty