Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Angelicalactone 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Methylfuran-2(3H)-one

L07602-14 L07602-14 L07602-22 L07602-22 L07602-06 L07602-06
AAAL07602-22EA 219.99 USD InStock
CAAAAL07602-22 AAAL07602-22 AAAL07602-14 CAAAAL07602-14 AAAL07602-06 CAAAAL07602-06
α-Angelicalactone 98%
α-Angelicalactone


Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 53…54 °C/10 mm
Melting Pt: 17…18 °C
Density: 1,093
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005375
CAS Number: 591-12-8
EINECS: 209-701-8
Merck Index: 12,00686
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty