Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-DL-phenylalanine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Amino-3-(2-fluorophenyl)propanoic acid

L07654-06 L07654-03 L07654-06 L07654-03
AAAL07654-03EA 35.19 USD InStock
CAAAAL07654-03 CAAAAL07654-06 AAAL07654-03 AAAL07654-06
2-Fluoro-DL-phenylalanine 98%
2-Fluoro-DL-phenylalanine


Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
Melting Pt: ca 245 °C (dec.)
Storage Temperature: Ambient
EINECS: 220-105-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty