Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-nitrocoumarin 98%

Supplier: Thermo Scientific Chemicals
Warning

L07908-03 L07908-06 L07908-03 L07908-06
AAAL07908-06EA 94.89 USD InStock
AAAL07908-06 AAAL07908-03 CAAAAL07908-06 CAAAAL07908-03
4-Chloro-3-nitrocoumarin 98%
4-Chloro-3-nitrocoumarin


Formula: C₉H₄ClNO₄
Melting Pt: ca 162 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051670
CAS Number: 38464-20-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty