Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Linoleic acid 95%

Supplier: Thermo Scientific Chemicals
Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid9Z,12Z-Octadecadienoic acidcis,cis-9,12-Octadecadienoic acid

L07949-14 L07949-06 L07949-14 L07949-22 L07949-06 L07949-22
AAAL07949-14EA 230.84 USD InStock
CAAAAL07949-22 AAAL07949-14 AAAL07949-06 CAAAAL07949-06 CAAAAL07949-14 AAAL07949-22
Linoleic acid 95%
Linoleic acid
Unsaturated omega-6 fatty acid


Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
Boiling Pt: 230 °C/16 mm
Melting Pt: –12 °C
Density: 0.902
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9
Merck Index: 13,05527
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty