Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-fluorobenzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Fluorobenzoic acid ethyl esterEthyl 3-fluorobenzoate

L07984-18 L07984-09 L07984-09 L07984-18
AAAL07984-09EA 36.69 USD InStock
AAAL07984-09 AAAL07984-18 CAAAAL07984-09 CAAAAL07984-18
Ethyl-3-fluorobenzoate 98+%
Ethyl 3-fluorobenzoate


Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 208 °C
Density: 1,136
Flash Pt: 82 °C(179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000336
CAS Number: 451-02-5
EINECS: 207-191-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty